Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202406.1 | NP_001189335.1 | 6 | 871 | ||
NM_001202407.1 | NP_001189336.1 | 6 | 881 | |||
NM_001202408.1 | NP_001189337.1 | 6 | 949 | |||
NM_001202409.1 | NP_001189338.1 | 7 | 1171 | |||
NM_001202410.1 | NP_001189339.1 | 7 | 1019 | |||
NM_001202411.1 | NP_001189340.1 | 7 | 1048 | |||
NM_001202412.1 | NP_001189341.1 | 7 | 734 | |||
NM_032497.2 | NP_115886.1 | 7 | 934 | |||
GenBank mRNA | AB075488.1 | - | 2 | 234 | ||
AK027700.1 | - | 7 | 612 | |||
AK056489.1 | - | 6 | 446 | |||
AK092348.1 | - | 5 | 968 | |||
AK298169.1 | - | 4 | 225 | |||
AK308517.1 | - | 6 | 542 | |||
AK310020.1 | - | 7 | 746 | |||
AK310117.1 | - | 6 | 1037 | |||
AK310781.1 | - | 4 | 866 | |||
BC006436.1 | - | 6 | 919 | 3957973 | ||
BP417021.1 | - | 6 | 461 | |||
CF597362.1 | - | 7 | 541 | |||
DA774704.1 | - | 5 | 348 |