Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STAT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012448.3 | NP_036580.2 | 2 | 264 | ||
XM_005257626.4 | XP_005257683.1 | 2 | 318 | |||
XM_017024977.1 | XP_016880466.1 | 2 | 240 | |||
GenBank mRNA | AB208920.1 | - | 2 | 243 | ||
AK308163.1 | - | 2 | 152 | |||
BC020868.1 | - | 2 | 264 | |||
BC065227.1 | - | 2 | 264 | 5553426 | ||
U47686.1 | - | 2 | 241 | |||
U48730.2 | - | 1 | 106 |