Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: JAGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032492.3 | NP_115881.3 | 2 | 546 | ||
GenBank mRNA | AF212230.1 | - | 2 | 486 | ||
AK027508.1 | - | 2 | 492 | |||
AK074760.1 | - | 2 | 545 | |||
AK225756.1 | - | 2 | 528 | |||
BC032101.2 | - | 2 | 526 | 4554225 | ||
BP262753.1 | - | 2 | 561 |