Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: JAGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032492.3 | NP_115881.3 | 2 | 851 | ||
GenBank mRNA | AF212230.1 | - | 2 | 792 | ||
AK027508.1 | - | 2 | 797 | |||
AK074760.1 | - | 2 | 850 | |||
AK225756.1 | - | 2 | 833 | |||
BC032101.2 | - | 2 | 831 | 4554225 |