Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321209.1 | NP_001308138.1 | 5 | 423 | ||
NM_032574.3 | NP_115963.1 | 5 | 563 | |||
NR_135585.1 | 5 | 416 | ||||
NR_135587.1 | 4 | 280 | ||||
GenBank mRNA | AA442557.1 | - | 4 | 211 | ||
AF226998.1 | - | 5 | 482 | |||
AV725116.1 | - | 5 | 354 | |||
BC015970.1 | - | 5 | 385 | 4081996 | ||
BI752326.1 | - | 5 | 384 | |||
BQ276899.1 | - | 423 |