Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032728.3 | NP_116117.3 | 1 | 738 | ||
XM_005272230.3 | XP_005272287.1 | 1 | 750 | |||
XM_005272231.1 | XP_005272288.1 | 1 | 750 | |||
GenBank mRNA | AK027568.1 | - | 1 | 692 | ||
AK075207.1 | - | 1 | 738 | |||
AK315502.1 | - | 1 | 567 | |||
BC006362.2 | - | 1 | 695 | 4129897 |