Search Thermo Fisher Scientific
Gene Symbol: PHF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032758.3 | NP_116147.1 | 4 | 573 | ||
GenBank mRNA | BC007321.2 | - | 4 | 556 | 2989046 | |
BC075808.1 | - | 4 | 541 | 5574722 | ||
CR456398.1 | - | 4 | 543 |