Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032777.9 | NP_116166.9 | 3 | 741 | ||
XM_005273471.3 | XP_005273528.1 | 3 | 741 | |||
XM_011544481.2 | XP_011542783.1 | 3 | 741 | |||
XM_011544482.2 | XP_011542784.1 | 2 | 669 | |||
XM_011544483.2 | XP_011542785.1 | 3 | 741 | |||
GenBank mRNA | AB040964.2 | - | 3 | 718 | ||
AF378755.1 | - | 3 | 368 | |||
AK027296.1 | - | 3 | 741 | |||
AK074099.2 | - | 3 | 272 | |||
AY090637.1 | - | 3 | 703 | |||
AY181242.1 | - | 3 | 334 | |||
BC146774.1 | - | 3 | 718 | 9007223 | ||
KU257672.1 | - | 3 | 253 |
Set Membership: |
![]() |