Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256421.1 | NP_001243350.1 | 9, 10 | 1137 | ||
NM_001256422.1 | NP_001243351.1 | 9, 10 | 1141 | |||
NM_032717.4 | NP_116106.2 | 8, 9 | 1224 | |||
XM_011532383.1 | XP_011530685.1 | 7, 8 | 1030 | |||
XM_011532384.2 | XP_011530686.1 | 9, 10 | 3225 | |||
XM_017008780.1 | XP_016864269.1 | 9, 10 | 1293 | |||
XM_017008781.1 | XP_016864270.1 | 9, 10 | 1300 | |||
GenBank mRNA | AB075872.1 | - | 5, 6 | 422 | ||
AK055749.1 | - | 9, 10 | 1136 | |||
AK223543.1 | - | 9, 10 | 1108 | |||
AY358100.1 | - | 8, 9 | 1041 | |||
BC073136.1 | - | 3, 4 | 200 | |||
BC090956.1 | - | 8, 9 | 1130 | 30387839 | ||
DQ324782.1 | - | 8, 9 | 921 | |||
DQ345298.1 | - | 8, 9 | 1221 |