Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032777.9 | NP_116166.9 | 18 | 3071 | ||
XM_011544481.2 | XP_011542783.1 | 18 | 3140 | |||
XM_011544482.2 | XP_011542784.1 | 17 | 3068 | |||
GenBank mRNA | AB040964.2 | - | 15 | 2397 | ||
AF378755.1 | - | 18 | 2698 | |||
AK074099.2 | - | 18 | 2602 | |||
AL832524.1 | - | 2 | 138 | |||
AL832959.1 | - | 11 | 1741 | |||
AY181242.1 | - | 18 | 2664 | |||
BC146774.1 | - | 15 | 2397 | 9007223 |
Set Membership: |
![]() |