Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317895.1 | NP_001304824.1 | 4 | 1196 | ||
NM_032789.4 | NP_116178.2 | 5 | 1072 | |||
NR_134234.1 | 5 | 1072 | ||||
XM_011517336.2 | XP_011515638.1 | 5 | 1039 | |||
GenBank mRNA | AK027370.1 | - | 5 | 1029 | ||
AK128537.1 | - | 6 | 887 | |||
AK222914.1 | - | 5 | 1055 | |||
AK303317.1 | - | 4 | 1121 | |||
AK308657.1 | - | 4 | 876 | |||
AK309251.1 | - | 4 | 776 | |||
BC020378.1 | - | 5 | 1047 | |||
BC030549.1 | - | 5 | 1046 | |||
BC132961.1 | - | 5 | 1023 | 40146983 | ||
BC136542.1 | - | 5 | 1023 | 9020532 | ||
BC144235.1 | - | 5 | 1023 |