Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079669.2 | NP_001073137.1 | 16, 17 | 2261 | ||
NM_001286453.1 | NP_001273382.1 | 14, 15 | 1928 | |||
NM_032813.3 | NP_116202.2 | 17, 18 | 2471 | |||
XM_011521121.2 | XP_011519423.1 | 18, 19 | 2623 | |||
XM_011521122.2 | XP_011519424.1 | 18, 19 | 2539 | |||
XM_011521123.2 | XP_011519425.1 | 18, 19 | 2700 | |||
XM_011521124.2 | XP_011519426.1 | 17, 18 | 2328 | |||
XM_017020796.1 | XP_016876285.1 | 17, 18 | 2526 | |||
XM_017020797.1 | XP_016876286.1 | 16, 17 | 2265 | |||
XM_017020798.1 | XP_016876287.1 | 16, 17 | 2361 | |||
XM_017020799.1 | XP_016876288.1 | 15, 16 | 2046 | |||
GenBank mRNA | AF339792.1 | - | 14, 15 | 1843 | ||
AK027530.1 | - | 11, 12 | 1354 | |||
AK056409.1 | - | 17, 18 | 2330 | |||
AK290345.1 | - | 16, 17 | 2121 | |||
AK299859.1 | - | 14, 15 | 1767 | |||
BX647956.1 | - | 16, 17 | 2152 | |||
BX648008.1 | - | 16, 17 | 2105 | |||
DA368450.1 | - | 4, 5 | 425 |