Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032836.2 | NP_116225.2 | 2 | 272 | ||
XM_005259352.4 | XP_005259409.1 | 2 | 1297 | |||
GenBank mRNA | AK027674.1 | - | 2 | 272 | ||
AK160385.1 | - | 1 | 4285 | |||
BC020368.1 | - | 1 | 574 | |||
BC132777.1 | - | 1 | 233 | 40146799 | ||
BC136512.1 | - | 1 | 231 | 9020502 |