Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301163.1 | NP_001288092.1 | 2 | 203 | ||
NM_032842.3 | NP_116231.2 | 2 | 203 | |||
XM_005250655.1 | XP_005250712.1 | 2 | 493 | |||
XM_011516646.2 | XP_011514948.1 | 2 | 343 | |||
GenBank mRNA | AK027678.1 | - | 2 | 108 | ||
AK027709.1 | - | 2 | 113 | |||
AK074732.1 | - | 2 | 136 | |||
BC045618.1 | - | 2 | 111 | 5294911 | ||
BC057784.1 | - | 2 | 108 | 5313685 | ||
BU177118.1 | - | 2 | 148 | |||
DB021198.1 | - | 2 | 203 |