Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032873.4 | NP_116262.2 | 2 | 555 | ||
XM_005271712.3 | XP_005271769.1 | 2 | 2801 | |||
XM_011543041.2 | XP_011541343.1 | 2 | 189 | |||
XM_011543042.1 | XP_011541344.1 | 2 | 555 | |||
GenBank mRNA | AB075839.1 | - | 2 | 407 | ||
AK075203.1 | - | 2 | 523 | |||
BC007541.1 | - | 2 | 509 | 2958242 | ||
DA197468.1 | - | 2 | 555 |