Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC22C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201572.1 | NP_001188501.1 | 2 | 257 | ||
NM_001201584.1 | NP_001188513.1 | 2 | 319 | |||
NM_004206.3 | NP_004197.1 | 2 | 319 | |||
NM_032970.3 | NP_116752.1 | 2 | 319 | |||
XM_005265533.1 | XP_005265590.1 | 2 | 277 | |||
XM_005265534.1 | XP_005265591.1 | 2 | 269 | |||
XM_005265535.1 | XP_005265592.1 | 2 | 235 | |||
XM_011534212.1 | XP_011532514.1 | 2 | 171 | |||
XM_017007438.1 | XP_016862927.1 | 2 | 277 | |||
XM_017007439.1 | XP_016862928.1 | 2 | 268 | |||
GenBank mRNA | AF039568.1 | - | 1 | 109 | ||
AK074576.1 | - | 2 | 319 | |||
AK292856.1 | - | 2 | 253 | |||
AK293133.1 | - | 2 | 253 | |||
AY359041.1 | - | 2 | 245 | |||
BC006178.1 | - | 2 | 224 | 4054516 | ||
BC018437.1 | - | 2 | 227 | 3445481 | ||
BX377531.2 | - | 2 | 257 | |||
DA434403.1 | - | 2 | 255 | |||
KU178463.1 | - | 1 | 109 | |||
KU178464.1 | - | 1 | 109 | |||
KU178465.1 | - | 1 | 109 |