Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300910.1 | NP_001287839.1 | 6 | 1305 | ||
NM_033025.5 | NP_149014.3 | 6 | 1506 | |||
GenBank mRNA | AK123686.1 | - | 6 | 1467 | ||
AK128870.1 | - | 3 | 2049 | |||
AK315453.1 | - | 6 | 1264 | |||
BC018942.2 | - | 2 | 70 | |||
BC029926.1 | - | 6 | 1247 | 5106728 |