Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HSH2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291274.1 | NP_001278203.1 | 7 | 1480 | ||
NM_032855.3 | NP_116244.1 | 7 | 947 | |||
NR_111903.1 | 7 | 954 | ||||
NR_111904.1 | 6 | 1348 | ||||
XM_011528386.2 | XP_011526688.1 | 5 | 538 | |||
XM_011528387.2 | XP_011526689.1 | 5 | 540 | |||
XM_011528388.2 | XP_011526690.1 | 6 | 618 | |||
XM_011528389.2 | XP_011526691.1 | 5 | 568 | |||
XM_011528390.1 | XP_011526692.1 | 5 | 533 | |||
XM_017027402.1 | XP_016882891.1 | 5 | 533 | |||
XM_017027403.1 | XP_016882892.1 | 5 | 533 | |||
XM_017027404.1 | XP_016882893.1 | 5 | 539 | |||
GenBank mRNA | AK027792.1 | - | 5 | 527 | ||
AK131222.1 | - | 5 | 1060 | |||
AK298682.1 | - | 4 | 891 | |||
AK300027.1 | - | 4 | 904 | |||
AK307563.1 | - | 5 | 497 | |||
AY319652.1 | - | 7 | 947 | |||
BC025237.2 | - | 4 | 447 | 4853087 |