Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281504.1 | NP_001268433.1 | 9 | 1484 | ||
NM_032885.5 | NP_116274.3 | 9 | 1410 | |||
NR_104024.1 | 9 | 1366 | ||||
NR_104025.1 | 8 | 1133 | ||||
XM_006722927.1 | XP_006722990.1 | 7 | 1065 | |||
GenBank mRNA | AJ312332.1 | - | 9 | 1226 | ||
AK223383.1 | - | 9 | 1335 | |||
AK294676.1 | - | 9 | 1484 | |||
AK314712.1 | - | 9 | 1345 | |||
BC007639.1 | - | 8 | 1056 | |||
BC016845.1 | - | 4 | 368 | |||
BC068992.1 | - | 9 | 1386 | 6578102 |