Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033102.2 | NP_149093.1 | 2 | 494 | ||
XM_005245556.3 | XP_005245613.1 | 2 | 660 | |||
XM_005245557.4 | XP_005245614.1 | 2 | 586 | |||
XM_005245560.2 | XP_005245617.1 | 2 | 494 | |||
XM_017002638.1 | XP_016858127.1 | 2 | 1033 | |||
GenBank mRNA | AK290364.1 | - | 2 | 495 | ||
AK313479.1 | - | 2 | 485 | |||
AY033593.1 | - | 2 | 438 | |||
BC050416.2 | - | 2 | 402 | 6198823 |