Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033051.3 | NP_149040.3 | 2, 3 | 1440 | ||
GenBank mRNA | AF242557.1 | - | 2, 3 | 1440 | ||
AK055571.1 | - | 3, 4 | 1380 | |||
AK314414.1 | - | 2, 3 | 1347 | |||
BC048285.1 | - | 2, 3 | 1292 | 5213610 |