Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027123.1 | XP_016882612.1 | 7 | 989 | ||
XM_017027124.1 | XP_016882613.1 | 7 | 991 | |||
XM_017027125.1 | XP_016882614.1 | 7 | 994 | |||
XM_017027126.1 | XP_016882615.1 | 7 | 1084 | |||
XM_017027127.1 | XP_016882616.1 | 7 | 949 | |||
XM_017027128.1 | XP_016882617.1 | 7 | 1154 | |||
GenBank mRNA | AK057404.1 | - | 7 | 976 | ||
AK301822.1 | - | 7 | 975 |