Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033103.4 | NP_149094.3 | 4, 5 | 464 | ||
GenBank mRNA | AF268032.1 | - | 4, 5 | 425 | ||
AF423421.1 | - | 4, 5 | 398 | |||
AJ347750.1 | - | 4, 5 | 398 | |||
AK098246.1 | - | 4, 5 | 434 | |||
AK126506.1 | - | 2, 3 | 857 | |||
AK315105.1 | - | 4, 5 | 434 | |||
AL831950.1 | - | 3, 4 | 312 | |||
BC036447.2 | - | 4, 5 | 434 | 4830913 | ||
DC348657.1 | - | 4, 5 | 465 |