Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256430.1 | NP_001243359.1 | 6 | 1513 | ||
NM_033104.3 | NP_149095.2 | 5 | 1409 | |||
XM_006720279.1 | XP_006720342.1 | 4 | 1298 | |||
XM_011537240.1 | XP_011535542.1 | 5 | 1469 | |||
XM_011537241.1 | XP_011535543.1 | 5 | 1469 | |||
XM_011537242.1 | XP_011535544.1 | 5 | 1469 | |||
GenBank mRNA | AB208948.1 | - | 6 | 1513 | ||
AF255309.1 | - | 1 | 608 | |||
AF380833.1 | - | 4 | 1143 | |||
AF449430.1 | - | 4 | 1301 | |||
BC069359.1 | - | 4 | 1281 | 7262416 | ||
BC069389.1 | - | 4 | 1275 | 7262428 | ||
BC069502.1 | - | 4 | 1281 | 7262452 | ||
BC069587.1 | - | 4 | 1282 | |||
BC117493.1 | - | 4 | 1211 | 40126044 | ||
BC117495.1 | - | 4 | 1211 | 40126046 |