Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033256.2 | NP_150281.1 | 2, 3 | 542 | ||
GenBank mRNA | AB056508.1 | - | 2, 3 | 483 | ||
AF123763.1 | - | 2, 3 | 333 | |||
AY050670.1 | - | 2, 3 | 327 | |||
BC021089.1 | - | 2, 3 | 307 | 5019808 |