Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146172.1 | NP_001139644.1 | 2 | 272 | ||
NM_001146173.1 | NP_001139645.1 | 2 | 272 | |||
NM_033438.3 | NP_254273.2 | 2 | 272 | |||
XM_017002756.1 | XP_016858245.1 | 3 | 1828 | |||
GenBank mRNA | AF275725.1 | - | 2 | 239 | ||
AY034613.1 | - | 2 | 272 | |||
AY358801.1 | - | 2 | 235 | |||
BC070213.1 | - | 2 | 200 | |||
BC074754.2 | - | 2 | 190 | 30915213 |