Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168299.1 | NP_001161771.1 | 4 | 445 | ||
NM_001168300.1 | NP_001161772.1 | 3 | 693 | |||
NM_001168301.1 | NP_001161773.1 | 4 | 343 | |||
NM_001168302.1 | NP_001161774.1 | 4 | 637 | |||
NM_001168303.1 | NP_001161775.1 | 3 | 563 | |||
NM_033495.3 | NP_277030.2 | 3 | 1213 | |||
XM_011531409.2 | XP_011529711.1 | 5 | 1105 | |||
XM_011531410.2 | XP_011529712.1 | 5 | 405 | |||
XM_011531411.1 | XP_011529713.1 | 4 | 784 | |||
XM_017029950.1 | XP_016885439.1 | 3 | 1474 | |||
GenBank mRNA | AB037730.1 | - | 2 | 2572 | ||
AK122724.1 | - | 3 | 280 | |||
AK125356.1 | - | 3 | 1213 | |||
AK296324.1 | - | 4 | 352 | |||
AK297523.1 | - | 3 | 385 | |||
AK299257.1 | - | 3 | 457 | |||
AK302713.1 | - | 4 | 343 | |||
AK316509.1 | - | 4 | 597 | |||
AL591986.1 | - | 3 | 360 | |||
BC064576.1 | - | 3 | 365 | 5727768 | ||
DC354391.1 | - | 4 | 445 |