Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017919.1 | NP_001017919.1 | 5, 6 | 977 | ||
NM_033544.2 | NP_291022.2 | 6, 7 | 1061 | |||
GenBank mRNA | BC064606.1 | - | 3, 4 | 988 | ||
BC094739.1 | - | 5, 6 | 1264 | |||
BC113826.1 | - | 3, 4 | 385 | |||
BC140708.1 | - | 5, 6 | 975 | 9021576 | ||
BC140709.1 | - | 5, 6 | 975 | 9021577 | ||
BU850147.1 | - | 4, 5 | 778 |