Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322923.1 | NP_001309852.1 | 5 | 1241 | ||
NM_001322924.1 | NP_001309853.1 | 4 | 1169 | |||
NM_052862.3 | NP_443094.3 | 6 | 1287 | |||
GenBank mRNA | AF545852.1 | - | 6 | 1431 | ||
AK025425.1 | - | 1 | 626 | |||
AL137762.1 | - | 6 | 1208 | |||
AL833211.1 | - | 4 | 977 | |||
AY530954.1 | - | 5 | 1100 | |||
BC013186.1 | - | 1 | 336 | |||
BC072399.1 | - | 1 | 579 | |||
BC098426.1 | - | 1 | 452 | |||
BC101536.1 | - | 6 | 1124 | 8069042 | ||
BC101562.1 | - | 6 | 1124 | 8069068 | ||
BC143428.1 | - | 5 | 1034 | 9051936 |