Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102610.1 | NP_001096080.1 | 3 | 405 | ||
NM_052903.4 | NP_443135.3 | 3 | 405 | |||
XM_005272540.2 | XP_005272597.1 | 3 | 309 | |||
XM_011543868.1 | XP_011542170.1 | 3 | 309 | |||
XM_011543869.2 | XP_011542171.1 | 3 | 309 | |||
XM_011543870.1 | XP_011542172.1 | 3 | 309 | |||
XM_017021890.1 | XP_016877379.1 | 3 | 309 | |||
XM_017021891.1 | XP_016877380.1 | 3 | 309 | |||
XM_017021892.1 | XP_016877381.1 | 3 | 309 | |||
XM_017021893.1 | XP_016877382.1 | 3 | 309 | |||
XM_017021895.1 | XP_016877384.1 | 3 | 329 | |||
XM_017021899.1 | XP_016877388.1 | 3 | 301 | |||
XM_017021900.1 | XP_016877389.1 | 3 | 273 | |||
GenBank mRNA | AB067486.2 | - | 3 | 319 | ||
AF272884.1 | - | 3 | 329 | |||
AK056416.1 | - | 3 | 304 | |||
AK289859.1 | - | 3 | 304 | |||
AK310897.1 | - | 3 | 301 | |||
BC071560.1 | - | 3 | 301 | 5297149 | ||
BP362649.1 | - | 3 | 405 |