Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033426.2 | NP_219494.2 | 3 | 414 | ||
GenBank mRNA | AB051524.1 | - | 2 | 289 | ||
AK315124.1 | - | 3 | 391 | |||
AL834514.1 | - | 3 | 317 | |||
BC037300.2 | - | 3 | 334 | 4827858 |