Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRF3 SIRNA Assays ›
Gene Symbol:
IRF3
Gene Name:
interferon regulatory factor 3
Gene Aliases:
IIAE7
Chromosome Location:
Chr.19: 49659569 - 49665875 on Build GRCh38
UniGene ID:
Hs.289052
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
3549
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRF3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197122.1
NP_001184051.1
4, 5
669
NM_001197123.1
NP_001184052.1
4, 5
727
NM_001197124.1
NP_001184053.1
4, 5
669
NM_001197125.1
NP_001184054.1
3, 4
496
NM_001197126.1
NP_001184055.1
2, 3
324
NM_001197127.1
NP_001184056.1
3, 4
496
NM_001197128.1
NP_001184057.1
2, 3
324
NM_001571.5
NP_001562.1
4, 5
669
NR_045568.1
4, 5
816
XM_006723197.1
XP_006723260.1
4, 5
647
XM_006723198.1
XP_006723261.1
4, 5
583
XM_006723200.1
XP_006723263.1
3, 4
332
XM_006723201.1
XP_006723264.1
3, 4
405
XM_006723202.2
XP_006723265.1
4, 5
554
XM_017026766.1
XP_016882255.1
4, 5
638
XM_017026767.1
XP_016882256.1
4, 5
583
GenBank mRNA
AB102884.1
-
2, 3
244
AB102887.1
-
3, 4
416
AK057577.1
-
3, 4
1435
AK292027.1
-
4, 5
798
AK314421.1
-
4, 5
582
BC000660.2
-
4, 5
571
3347928
BC009395.2
-
4, 5
596
4135383
BC071721.1
-
3, 4
758
5494536
BP347627.1
-
3, 4
337
HM357932.1
-
4, 5
557
HM357934.1
-
2, 3
154
HM357935.1
-
3, 4
326
HM357937.1
-
3, 4
326
Z56281.1
-
4, 5
462
Back To Top
More Information
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
apoptotic process
cellular response to DNA damage stimulus
lipopolysaccharide-mediated signaling pathway
negative regulation of type I interferon production
positive regulation of type I interferon production
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
TRIF-dependent toll-like receptor signaling pathway
MDA-5 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to exogenous dsRNA
type I interferon biosynthetic process
positive regulation of cytokine secretion
defense response to virus
interferon-gamma-mediated signaling pathway
type I interferon signaling pathway
positive regulation of type I interferon-mediated signaling pathway
cellular response to dsRNA
macrophage apoptotic process
programmed necrotic cell death
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription cofactor activity
protein binding
identical protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-vrvhj:80/10.239.161.228:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0