Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC18B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052831.2 | NP_439896.1 | 3 | 355 | ||
XM_017010236.1 | XP_016865725.1 | 3 | 546 | |||
GenBank mRNA | AK124442.1 | - | 3 | 234 | ||
AK289918.1 | - | 3 | 374 | |||
AK293853.1 | - | 3 | 309 | |||
BC069570.1 | - | 3 | 275 | 7262298 |