Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163321.2 | NP_001156793.2 | 11 | 2642 | ||
NM_001163322.2 | NP_001156794.1 | 11 | 2583 | |||
NM_001163323.2 | NP_001156795.1 | 10 | 3072 | |||
NM_001271835.1 | NP_001258764.1 | 10 | 3072 | |||
NM_001271836.1 | NP_001258765.1 | 10 | 3424 | |||
NM_033626.3 | NP_296375.1 | 10 | 3013 | |||
GenBank mRNA | AK056290.1 | - | 1 | 1609 | ||
AK125424.1 | - | 1 | 2607 | |||
BC008769.2 | - | 10 | 2994 | 3139519 |