Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080791.2 | NP_001074260.1 | 4 | 1009 | ||
NM_001080792.2 | NP_001074261.1 | 4 | 799 | |||
NM_052849.3 | NP_443081.1 | 4 | 984 | |||
GenBank mRNA | AK092214.1 | - | 5 | 896 | ||
AL832032.2 | - | 4 | 695 | |||
BC001673.2 | - | 3 | 707 | 2823236 | ||
CD686743.1 | - | 2 | 424 |