Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278733.1 | NP_001265662.1 | 2 | 327 | ||
NM_054114.4 | NP_473455.2 | 6 | 698 | |||
NM_138810.3 | NP_620165.1 | 6 | 698 | |||
NM_152133.2 | NP_687034.1 | 5 | 531 | |||
GenBank mRNA | AF385429.1 | - | 6 | 698 | ||
AF385430.1 | - | 5 | 531 | |||
AK097090.1 | - | 6 | 564 | |||
BC015859.1 | - | 6 | 568 | |||
BC111731.1 | - | 6 | 554 | 40035199 | ||
DA819174.1 | - | 2 | 327 |