Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052924.2 | NP_443156.2 | 8 | 990 | ||
XM_005250772.3 | XP_005250829.1 | 9 | 1176 | |||
XM_005250773.3 | XP_005250830.1 | 9 | 1176 | |||
XM_006716502.3 | XP_006716565.1 | 9 | 1194 | |||
XM_011516817.2 | XP_011515119.1 | 9 | 1194 | |||
XM_011516818.1 | XP_011515120.1 | 9 | 1851 | |||
XM_011516819.2 | XP_011515121.1 | 8 | 1035 | |||
XM_011516820.2 | XP_011515122.1 | 8 | 1378 | |||
XM_011516821.2 | XP_011515123.1 | 8 | 1382 | |||
XM_011516822.2 | XP_011515124.1 | 8 | 1527 | |||
XM_011516823.2 | XP_011515125.1 | 9 | 1195 | |||
XM_017013013.1 | XP_016868502.1 | 8 | 1066 | |||
XM_017013014.1 | XP_016868503.1 | 9 | 1195 | |||
XM_017013015.1 | XP_016868504.1 | 8 | 1508 | |||
XM_017013016.1 | XP_016868505.1 | 9 | 1176 | |||
GenBank mRNA | AB067516.1 | - | 8 | 848 | ||
AY082588.1 | - | 8 | 857 | |||
BC025767.1 | - | 8 | 958 | 5211354 |