Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052950.3 | NP_443182.1 | 5 | 745 | ||
XM_011534914.1 | XP_011533216.1 | 5 | 636 | |||
XM_011534915.2 | XP_011533217.1 | 4 | 825 | |||
XM_017020386.1 | XP_016875875.1 | 5 | 6549 | |||
GenBank mRNA | AF411978.1 | - | 5 | 405 | ||
AK058149.1 | - | 3 | 191 | |||
AK309815.1 | - | 5 | 598 | |||
BC014004.2 | - | 5 | 585 | 3842589 |