Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_079420.2 | NP_524144.1 | 3 | 305 | ||
NM_079422.2 | NP_524146.1 | 3 | 115 | |||
GenBank mRNA | AK311892.1 | - | 3 | 115 | ||
AK311942.1 | - | 3 | 326 | |||
BC005318.1 | - | 3 | 115 | 3933998 | ||
DA898482.1 | - | 3 | 331 |