Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080282.3 | NP_525021.3 | 5 | 1067 | ||
GenBank mRNA | AK310858.1 | - | 4 | 597 | ||
AL832004.2 | - | 4 | 575 | |||
AY028900.1 | - | 5 | 1067 | |||
AY247065.1 | - | 5 | 1098 | |||
BC136460.1 | - | 2 | 272 | |||
BC144124.1 | - | 2 | 272 |