Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080284.2 | NP_525023.2 | 2 | 257 | ||
XM_017024404.1 | XP_016879893.1 | 2 | 569 | |||
XM_017024405.1 | XP_016879894.1 | 2 | 569 | |||
GenBank mRNA | AF373290.1 | - | 1 | 85 | ||
AK308431.1 | - | 2 | 277 | |||
AY028898.1 | - | 2 | 257 | |||
BC070125.1 | - | 2 | 272 | |||
BC125231.1 | - | 2 | 256 | |||
BC125232.1 | - | 2 | 256 |