Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080675.3 | NP_542406.2 | 4 | 359 | ||
XM_011528573.1 | XP_011526875.1 | 5 | 443 | |||
XM_011528574.1 | XP_011526876.1 | 3 | 299 | |||
XM_011528576.1 | XP_011526878.1 | 5 | 444 | |||
GenBank mRNA | AF401350.1 | - | 4 | 359 | ||
AK097733.1 | - | 2 | 107 | |||
BC026118.2 | - | 4 | 359 | 4824412 | ||
BC029528.1 | - | 4 | 352 | 5271065 |