Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 7, 8 | 1487 | ||
NM_080616.4 | NP_542183.2 | 4, 5 | 826 | |||
XM_005260282.4 | XP_005260339.1 | 7, 8 | 1773 | |||
XM_005260283.4 | XP_005260340.1 | 8, 9 | 1797 | |||
XM_005260284.4 | XP_005260341.1 | 7, 8 | 1072 | |||
XM_005260285.4 | XP_005260342.2 | 7, 8 | 1777 | |||
XM_005260286.4 | XP_005260343.1 | 7, 8 | 1697 | |||
XM_005260288.2 | XP_005260345.1 | 4, 5 | 874 | |||
XM_005260289.4 | XP_005260346.1 | 4, 5 | 825 | |||
XM_006723702.3 | XP_006723765.1 | 8, 9 | 1164 | |||
XM_006723703.3 | XP_006723766.1 | 7, 8 | 1076 | |||
XM_011528562.2 | XP_011526864.1 | 7, 8 | 1481 | |||
XM_011528563.1 | XP_011526865.1 | 7, 8 | 1481 | |||
XM_017027669.1 | XP_016883158.1 | 9, 10 | 1818 | |||
XM_017027670.1 | XP_016883159.1 | 4, 5 | 729 | |||
GenBank mRNA | AK056286.1 | - | 4, 5 | 629 | ||
BC065370.1 | - | 3, 4 | 598 | 6261963 |