Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080650.3 | NP_542381.1 | 5 | 465 | ||
XM_011522160.1 | XP_011520462.1 | 5 | 484 | |||
XM_017022707.1 | XP_016878196.1 | 5 | 484 | |||
XM_017022708.1 | XP_016878197.1 | 5 | 484 | |||
XM_017022709.1 | XP_016878198.1 | 5 | 484 | |||
GenBank mRNA | AK094007.1 | - | 5 | 425 | ||
BC008485.2 | - | 4 | 381 | |||
BC066652.1 | - | 5 | 412 | 6066017 | ||
DB250468.1 | - | 5 | 465 |