Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080742.2 | NP_542780.1 | 2, 3 | 1400 | ||
GenBank mRNA | AB075843.1 | - | 2, 3 | 1353 | ||
AK055248.1 | - | 2, 3 | 276 | |||
AK290018.1 | - | 2, 3 | 300 | |||
AY070019.1 | - | 2, 3 | 744 | |||
BC101163.2 | - | 3, 4 | 384 | |||
BC101165.2 | - | 3, 4 | 393 | |||
BC113995.1 | - | 2, 3 | 528 | 40004155 |