Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080864.3 | NP_543140.1 | 1, 2 | 248 | ||
GenBank mRNA | AB076563.1 | - | 1, 2 | 191 | ||
AF447451.1 | - | 1, 2 | 191 | |||
AY358181.1 | - | 2, 3 | 1031 | |||
BC140935.1 | - | 1, 2 | 248 | 9056747 |