Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080874.3 | NP_543150.1 | 1, 2 | 318 | ||
XM_005262759.1 | XP_005262816.1 | 3, 4 | 747 | |||
GenBank mRNA | AY057053.1 | - | 1, 2 | 204 | ||
BC065710.1 | - | 1, 2 | 225 | 4272258 | ||
BX647857.1 | - | 1, 2 | 319 |