Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130775.2 | NP_570131.1 | 5 | 383 | ||
XM_017029320.1 | XP_016884809.1 | 6 | 616 | |||
GenBank mRNA | BC069129.1 | - | 5 | 384 | 7216985 | |
BC103962.1 | - | 5 | 837 | 40014176 | ||
BC103963.1 | - | 4 | 834 | |||
BC103964.1 | - | 4 | 834 |