Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324081.1 | NP_001311010.1 | 7 | 998 | ||
NM_080861.3 | NP_543137.2 | 7 | 1020 | |||
XM_005255673.4 | XP_005255730.1 | 7 | 1210 | |||
GenBank mRNA | AF403028.1 | - | 6 | 929 | ||
AK057850.1 | - | 778 | ||||
AK074190.1 | - | 7 | 984 | |||
BC007588.1 | - | 7 | 966 | 3342825 | ||
BC024244.1 | - | 7 | 984 | 4798873 | ||
BC035065.1 | - | 7 | 977 | |||
BC041897.1 | - | 6 | 1794 | 5298729 | ||
BC047441.1 | - | 7 | 962 | 5303706 | ||
BC065515.1 | - | 4 | 1263 | |||
BQ930544.1 | - | 2 | 302 |