Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ASB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080868.2 | NP_543144.1 | 3 | 942 | ||
NR_026546.1 | 3 | 880 | ||||
XM_006710345.3 | XP_006710408.1 | 4 | 774 | |||
XM_006710346.3 | XP_006710409.1 | 4 | 701 | |||
GenBank mRNA | AF429968.1 | - | 3 | 919 | ||
AK098606.1 | - | 3 | 942 | |||
BC022517.1 | - | 3 | 813 | |||
BC036573.2 | - | 3 | 920 | 5297837 |